missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaMKK beta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521929
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: PC-3. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CaMKK beta Polyclonal antibody specifically detects CaMKK beta in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CaMKK beta | |
Polyclonal | |
Unconjugated | |
CAMKK2 | |
6330570N16Rik; AW061083; Ca+/Calmodulin-dependent protein kinase kinase beta (CaM-kinase kinase beta); calcium/calmodulin dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase 2 beta; calcium/calmodulin-dependent protein kinase beta; calcium/calmodulin-dependent protein kinase kinase 2; calcium/calmodulin-dependent protein kinase kinase 2, beta; calcium/calmodulin-dependent protein kinase kinase beta; caM-kinase kinase 2; CaM-kinase kinase beta; CAMKK; caMKK 2; caM-KK 2; caMKK beta; CaM-KK beta; CAMKK beta protein; CAMKK2; CAMKKB; KIAA0787; MGC15254; mKIAA0787 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10645 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.73 mg/mL | |
PBS with 20% glycerol and 0.01% thimerosal, 0.025% ProClin 300; pH 7 | |
Q96RR4 | |
CAMKK2 | |
Synthetic peptide corresponding to a region within amino acids 1 and 498 of CaMKK beta | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |