Learn More
Calprotectin Monoclonal Antibody (27E10), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA140314
Code : LD
Additional Details : Weight : 0.10000kg
Description
MA1-40314 detects Calprotectin from human and non-human primate samples. Does not cross react with mouse. MA1-40314 has been successfully used in ELISA, immunohistochemistry (frozen), flow cytometry, immunofluorescence, immunoprecipitation and Western blot applications. For Western blot, detects the non-reduced protein at about 22 kDa. The MA1-40314 immunogen is proprietary.
Calprotectin Monoclonal antibody specifically detects Calprotectin in Human, Non-human Primate, Rhesus Monkey samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot.Specifications
Calprotectin | |
Monoclonal | |
0.1 mg/mL | |
PBS with 0.1% BSA and 0.02% sodium azide | |
P05109, P06702 | |
S100A8, S100A9 | |
Human blood monocytes | |
50 μg | |
Primary | |
Human, Non-human Primate, Rhesus Monkey | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
27E10 | |
Biotin | |
S100A9 | |
60B8Ag; AI323541; AW546964; B8Ag; BEE11; BEE22; CAGA; CAGB; calcium binding protein; S100 protein; calgranulin A; calgranulin B; calgranulin-A; Calgranulin-B; calprotectin; calprotectin L1H subunit; Calprotectin L1L subunit; CFAG; CGLA; CGLB; chemotactic cytokine CP-10; chemotactic S100 protein; CP-10; Cystic fibrosis antigen; EGK_01322; EGK_21438; GAGB; intracellular calcium-binding protein (MRP14); L1AG; Leukocyte L1 complex heavy chain; leukocyte L1 complex light chain; LIAG; LOC100735514; LOC532569 protein; MA387; MAC387; macrophage migration inhibitory factor-related protein-14; macrophage migration inhibitory factor-related protein-8; MIF; migration inhibitory factor-related protein 14; Migration inhibitory factor-related protein 8; Mrp14; MRP-14; MRP8; MRP-8; myeloid-related protein 14; Neutrophil cytosolic 23 kDa protein; neutrophil cytosolic 7 kDa protein; NIF; p14; p23; P7; p8; Pro-inflammatory S100 cytokine; protein S100-A8; Protein S100-A9; S100 calcium binding protein A8; | |
Mouse | |
Protein G | |
RUO | |
6279, 6280, 714690, 714740 | |
4° C | |
Liquid |