Learn More
Calmodulin Monoclonal Antibody (6D4), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA3918
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA3-918 detects calmodulin from bovine, chicken, Chlamydomonas, Dictyostelium, and rat samples. This antibody does not detect parvalbumin, tropinin, S-100, or myosin light chain kinase (MLCK). MA3-918 has been successfully used in Western blot, immunocytochemistry and ELISA procedures. By Western blot, this antibody detects a 17 kDa protein representing calmodulin from Dictyostelium cell lysate. Addition of EGTA to buffers completely inhibits antibody staining. Immunohistochemical staining of calmodulin in Dictyostelium cells with MA3-918 results in staining of the contractile vacuoles. The MA3-918 antigen is calmodulin purified from Dictyostelium discoideum.
Calmodulin Monoclonal antibody specifically detects Calmodulin in Bacteria, Bovine, Chicken, Human, Rat samples. It is validated for ELISA, Immunocytochemistry, Western Blot.Specifications
Calmodulin | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P05419, P0DP23, P0DP24, P0DP25, P0DP29, P0DP30, P0DP31, P62157 | |
CALM, Calm1, Calm2, CALM3 | |
Calmodulin purified from Dictyostelium discoideum. | |
100 μL | |
Primary | |
Bacteria, Bovine, Chicken, Human, Rat | |
Antibody | |
IgG1 |
ELISA, Immunocytochemistry, Western Blot | |
6D4 | |
Unconjugated | |
CALM3 | |
1500001E21Rik; 2310037J09Rik; AI256814; AI327027; AI461935; AI649108; AL024000; AL024017; AY314008; calcium binding gene, DD112; calcium-binding protein Dd112; CALM; CALM 1; CALM1; CALM2; Calm3; Calm4; Calm5; CALML2; Calmodulin; calmodulin 1; calmodulin 1 (phosphorylase kinase, delta); calmodulin 2; calmodulin 2 (phosphorylase kinase, delta); calmodulin 3; calmodulin 3 (phosphorylase kinase, delta); calmodulin 4; calmodulin 5; Calmodulin III; calmodulin-1; Calmodulin-2; calmodulin-3; Calmodulin-4; calmodulin-like; calmodulin-like protein; calmodulin-like protein (neoCaM); caM; CAM1; Cam2; CAM3; CaMA; Camb; CamC; CaMI; CAMII; CAMIII; CPVT4; DD112; DD132; DD312; epididymis secretory protein Li 72; HEL-S-72; hypothetical protein; LP7057 protein; LQT14; LQT15; MGC128137 protein; neo-calmodulin; NeoCaM; PHKD; PHKD2; PHKD3; phosphorylase kinase delta; phosphorylase kinase subunit delta; phosphorylase kinase, delta subunit; prepro-calmodulin 1; prepro-calmodulin 2; prepro-calmodulin 3; R75142 | |
Mouse | |
purified | |
RUO | |
100297344, 24242, 24244, 326597, 395855, 396523, 50663, 520277, 617095, 801, 805, 808 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |