missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CALML5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA531373
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A431. Predicted reactivity: Human (98%), Rhesus Monkey (83%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CALML5 Polyclonal antibody specifically detects CALML5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CALML5 | |
Polyclonal | |
Unconjugated | |
CALML5 | |
call5; Calm4; CALML5; calmodulin 4; calmodulin like 5; calmodulin-like 5; calmodulin-like protein 5; Calmodulin-like skin protein; CLSP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
51806 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9NZT1 | |
CALML5 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 131 of Human CALML5 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |