Learn More
Calcium Sensing Receptor Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1934A
Code : LE
Additional Details : Weight : 0.01000kg
Description
PA1-934A detects calcium sensing receptor (CaSR) from human and rat tissues. PA1-934A has been successfully used in Western blot and ICC/IF procedures. By Western blot, this antibody detects an ∽120 kDa protein representing CaSR from HEK393 cells and rat brain homogenate. The PA1-934A immunizing peptide corresponds to amino acid residues 12-27 from rat CaSR. This peptide (Cat.# PEP-009) is available for use in neutralization and control experiments.
Calcium Sensing Receptor Polyclonal antibody specifically detects Calcium Sensing Receptor in Human, Rat samples. It is validated for Immunocytochemistry, Western Blot.Specifications
Calcium Sensing Receptor | |
Polyclonal | |
Unconjugated | |
CASR | |
BoPCaR1; Ca sensing receptor; Ca(2+)-sensing receptor; calcium sensing receptor; calcium-sensing receptor; Calcium-sensing receptor (hypocalciuric hypercalcemia 1 severe neonatal hyperparathyroidism); Calcium-sensing receptor (hypocalciuric hypercalcemia 1, severe neonatal hyperparathyroidism); CaR; CaSR; CaSR antibody; cation sensing receptor; EIG8; Extracellular calcium-sensing receptor; FHH; FIH; G protein coupled receptor, family C, group 2, member A; G protein-coupled receptor, family C, group 2, member A; Gprc2a; HHC; HHC1; HYPOC1; LOW QUALITY PROTEIN: extracellular calcium-sensing receptor; NSHPT; parathyroid Ca(2+)-sensing receptor 1; parathyroid cell calcium-sensing receptor; Parathyroid cell calcium-sensing receptor 1; PCaR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
24247, 846 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P41180, P48442 | |
CASR | |
Synthetic peptide corresponding to residues A(12) L A W H S S A Y G P D Q R A Q(27) of rat CaSR. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |