missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CaV2.3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA112664
Code : LD
Additional Details : Weight : 0.01000kg
Description
CaV2.3 Polyclonal antibody specifically detects CaV2.3 in Human samples. It is validated for Western Blot.Specifications
CaV2.3 | |
Polyclonal | |
PBS with 0.08% sodium azide | |
Q15878 | |
Cacna1e | |
Synthetic peptide, conjugated to KLH, corresponding to the rat alpha1E calcium channel. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Cacna1e | |
A430040I15; alpha1E; BII; brain calcium channel II; CACH6; CACHA1E; CACNA1E; Cacnl1a6; calcium channel, L type, alpha-1 polypeptide, isoform 6; calcium channel, R type, alpha-1 polypeptide, isoform 6; calcium channel, voltage-dependent, L type, alpha 1E subunit; calcium channel, voltage-dependent, R type, alpha 1E subunit; calcium voltage-gated channel subunit alpha1 E; Cav2.3; Cchra1; gm139; RBE2; RBE-II; voltage gated calcium channel alpha1E subunit; voltage-dependent calcium channel alpha 1E subunit; voltage-dependent calcium channel alpha1-E subunit; voltage-dependent R-type calcium channel subunit alpha-1E; voltage-gated calcium channel alpha subunit Cav2.3; voltage-gated calcium channel subunit alpha Cav2.3 | |
Rabbit | |
Ammonium sulfate precipitation | |
RUO | |
777 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |