Learn More
CAD Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521352
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, C2C12. Predicted reactivity: Mouse (92%), Rat (92%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CAD Polyclonal antibody specifically detects CAD in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
CAD | |
Polyclonal | |
Unconjugated | |
CAD | |
2410008J01Rik; 38E.19; anon-WO2004063362.83; Aspartate carbamoyltransferase; AU018859; cad; CAD protein; CAD trifunctional protein; cad-PA; cad-PB; cad-PC; carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase; carbamoylphosphate synthetase 2/aspartate transcarbamylase/dihydroorotase; carbamyl phosphatate synthetase 2; caudal; cb456; cd; CDG1Z; CG1759; CG1759-PA; CG1759-PB; CG1759-PC; Cpad; Dihydroorotase; Dmel\CG1759; Dmel_CG1759; Glutamine-dependent carbamoyl-phosphate synthase; Homeotic protein caudal; LD29596p; multifunctional protein CAD; S67; si:dkey-221h15.3; wu:fc30c12; wu:fc33d01; wu:fc67g02 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
69719, 790 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 10% glycerol and 0.01% thimerosal; pH 7 | |
B2RQC6, P27708 | |
CAD | |
Recombinant fragment corresponding to a region within amino acids 1625 and 1930 of CAD (Uniprot ID#P27708) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |