missing translation for 'onlineSavingsMsg'
Learn More

C1qTNF9 Mouse anti-Human, Clone: 653533, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB6537

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13441406

  • £249.49 / 100µg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

C1qTNF9 Monoclonal antibody MAB6537 Specifically detects C1qTNF9 in Human samples. It is validated for Western Blotting
Specifications

Specifications

CTRP9/C1qTNF9
653533
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
C1QTNF9
Mouse myeloma cell line NS0-derived recombinant human CTRP9/C1qTNF9 Asn16-Pro333 Accession # P0C862
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
338872
Sterile PBS to a final concentration of 0.5 mg/mL.
Western Blot
LYOPH
Western Blot 1 ug/mL
P0C862
AQL1, C1q and tumor necrosis factor related protein 9, C1qTNF9, C1QTNF9AComplement C1q tumor necrosis factor-related protein 9, complement C1q tumor necrosis factor-related protein 9A, CTRP9, MGC48915
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CTRP9/C1qTNF9 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) C1qTNF1, rhCORS26/C1qTNF3, rhC1qTNF4, rhC1qTNF5, rhC1QL2 is observed.
Human
Videos

For Research Use Only