missing translation for 'onlineSavingsMsg'
Learn More

C1qTNF10/C1qL2 Rat anti-Mouse, Clone: 384921, R&D Systems™

Rat Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB4374

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13460536

  • £271.93 / 100µg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

C1qTNF10/C1qL2 Monoclonal antibody MAB4374 Specifically detects C1qTNF10/C1qL2 in Mouse samples. It is validated for Western Blotting
Specifications

Specifications

C1qTNF10/C1qL2
384921
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
C1QL2
Mouse myeloma cell line NS0-derived recombinant mouse C1qTNF10/C1qL2 His22-Asp287 Accession # Q8CFR0
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
165257
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q8CFR0
C1q and tumor necrosis factor related protein 10, C1q-domain containing protein, C1qL2, C1QTNF10, complement C1q-like protein 2, complement component 1, q subcomponent-like 2, CTRP10
Rat
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse C1qTNF10/C1qL2 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant human (rh) C1qTNF10 and no cross-reactivity with rhC1qTNF1, rhC1qTNF4, or rhC1qTNF5.
Mouse
Videos

For Research Use Only