Learn More
C1qTNF1 Mouse anti-Human, Clone: 395526, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3145
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469915
  • £288.60 / 100µg
Estimated Shipment
14-09-2020
Description
Description
C1qTNF1 Monoclonal antibody MAB3145 Specifically detects C1qTNF1 in Human samples. It is validated for Western Blotting
Specifications
Specifications
C1qTNF1/CTRP1
395526
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
C1QTNF1
Mouse myeloma cell line NS0-derived recombinant human CTRP1/C1qTNF1 Arg26-Pro281 Accession # Q9BXJ1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
114897
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9BXJ1
C1q and tumor necrosis factor related protein 1, C1qTNF1, CTRP1G protein-coupled receptor-interacting protein, FLJ90694, G protein coupled receptor interacting protein, GIPcomplement C1q tumor necrosis factor-related protein 1, ZSIG37
Mouse
IgG2b
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human CTRP1/C1qTNF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CTRP4, rhCTRP5, or rhC1qL2 is observed.