missing translation for 'onlineSavingsMsg'
Learn More

C1q R1/CD93 Mouse anti-Human, Clone: 273107, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB23791

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459395

  • £264.01 / 100µg


Estimated Shipment
30-10-2018
Add to basket

Description

Description

C1q R1/CD93 Monoclonal antibody MAB23791 Specifically detects C1q R1/CD93 in Human samples. It is validated for Flow Cytometry, Western Blotting
Specifications

Specifications

C1q R1/CD93
273107
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CD93
Mouse myeloma cell line NS0-derived recombinant human C1q R1/CD93, Ala24-Lys580, Accession # Q9NPY3
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
22918
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
LYOPH
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q9NPY3
AA4 Antigen, C1q receptor 1, C1q Rp, C1q/MBL/SPA receptor, C1qR(P), C1qR1, C1qRp, CD93, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Collectin Receptor, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Ly68, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Monoclonal
Detects human C1q R1/CD93 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse C1q R1 is observed.
Human
Videos

For Research Use Only