Learn More
C-Reactive Protein/CRP Rat anti-Mouse, Clone: 267917, R&D Systems™
Rat Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1829
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13448955
  • £376.32 / 500µg
Call for latest availability
+44 (0)1509 555 500
Description
Description
C-Reactive Protein/CRP Monoclonal antibody MAB1829 Specifically detects C-Reactive Protein/CRP in Mouse samples. It is validated for ELISA Capture
Specifications
Specifications
C-Reactive Protein/CRP
267917
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CRP
Mouse myeloma cell line NS0-derived recombinant mouse C-Reactive Protein Met1-Ser225 Accession # P14847
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
1401
Reconstitute at 0.5 mg/mL in sterile PBS.
ELISA
LYOPH
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P14847
C-reactive protein, C-reactive protein, pentraxin-related, CRP, MGC88244, pentraxin 1, PTX1MGC149895
Rat
IgG2a
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects mouse C-Reactive Protein in ELISAs.