missing translation for 'onlineSavingsMsg'
Learn More

c-Fos, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Brand:  Invitrogen PA118328

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11561992

  • £303.00 / 100µL


Estimated Shipment
27-06-2018
Add to basket

Description

Description

PA1-18328 detects c-FOS from rat, rabbit, and hamster samples.The sequence is 100% conserved in human, rat, mouse, hamster, and cat.PA1-18328 has been successfully used in immunohistochemistry and Western blot procedures.The PA1-18328 immunogen is a synthetic peptide conjugated to diphtheria toxoid residues M(2) F S G F N A D Y E A S S S R C(17), conjugated to diphtheria toxoid.After reconstitution keep aliquots at -20ºC for a higher stability, and at 4ºC with an appropriate antibacterial agent.Reconstitute with 100 ul of distilled water. Glycerol (1:1) may be added for additional stability.
Specifications

Specifications

c-Fos
Polyclonal
whole serum with no preservative
P12841
Cellular Oncogene Fos
Sheep
IgG
store at -20°C, Avoid Freeze/Thaw Cycles
Primary
100360880, 314322
ImmunohistochemistryþWestern Blot
Unconjugated
Fosb
Serum
Fos, FOSB
Synthetic peptide conjugated to diphtheria toxoid residues M(2) F S G F N A D Y E A S S S R C(17).
100μL
Antibody
Polyclonal
HamsterþRabbitþRat
Videos