missing translation for 'onlineSavingsMsg'
Learn More

BoNT-B Light Chain Sheep anti-C. botulinum, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF5420

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13459244

  • £384.13 / 100µg


Estimated Shipment
30-10-2018
Add to basket

Description

Description

BoNT-B Light Chain Polyclonal antibody AF5420 Specifically detects BoNT-B Light Chain in C. botulinum samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

BoNT-B Light Chain
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
P10844
boNT/B
E. coli-derived recombinant Clostridium BoNT-B Light Chain, Pro2-His428, Accession # P10844
Antigen Affinity-purified
RUO
Primary
6030102
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Polyclonal
Detects C. botulinum BoNT-B Light Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant Light Chains of BoNT-A, -E, or recombinant Heavy Chains of BoNT-C, or -D is observed.
C. botulinum
Videos

For Research Use Only