missing translation for 'onlineSavingsMsg'
Learn More

BoNT-A Light Chain Sheep anti-C. botulinum, Polyclonal, R&D Systems™

Sheep Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF4489

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448754

  • £344.84 / 100µg

Estimated Shipment date
from Supplier 09-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BoNT-A Light Chain Polyclonal antibody AF4489 Specifically detects BoNT-A Light Chain in C. botulinum samples. It is validated for Immunoprecipitation, Western Blotting
Specifications

Specifications

BoNT-A Light Chain
LYOPH
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL
Q45894
atx
E. coli-derived recombinant Clostridium BoNT-A Light Chain Met1-Phe425 Accession # Q45894
Immunogen affinity purified
RUO
Primary
5185061
Reconstitute at 0.2 mg/mL in sterile PBS.
Western Blot, Immunoprecipitation
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
Sheep
IgG
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects C. botulinum BoNTA Light Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant C. botulinum BoNTA Heavy Chain is observed.
C. botulinum
Videos
Documents

For Research Use Only