Learn More
ABCB11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527742
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat. Predicted reactivity: Mouse (85%), Rat (87%), Dog (84%), Cat (87%), Rabbit (85%), Bovine (88%), Guinea pig (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ABCB11 Polyclonal antibody specifically detects ABCB11 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
ABCB11 | |
Polyclonal | |
Unconjugated | |
ABCB11 | |
ABC member 16, MDR/TAP subfamily; ABC16; Abcb11; ATP binding cassette subfamily B member 11; ATP-binding cassette sub-family B member 11; ATP-binding cassette, subfamily B (MDR/TAP), member 11; ATP-binding cassette, sub-family B (MDR/TAP), member 11; ATP-binding cassette, sub-family B, member 11; bile salt export pump; BRIC2; Bsep; Lith1; PFIC2; PFIC-2; PGY4; progressive familial intrahepatic cholestasis 2; sister of P-glycoprotein; sister p-glycoprotein; Spgp | |
Rabbit | |
Protein A | |
RUO | |
8647 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.17 mg/mL | |
1 XPBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
O95342 | |
ABCB11 | |
Recombinant fragment corresponding to a region within amino acids 474 and 745 of Human BSEP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |