missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seipin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA523621
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with bovine based on sequence homology.
Seipin Polyclonal antibody specifically detects Seipin in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Seipin | |
Polyclonal | |
Unconjugated | |
BSCL2 | |
2900097C17Rik; AI046355; Berardinelli-Seip congenital lipodystrophy 2 (seipin); Berardinelli-Seip congenital lipodystrophy 2 homolog (seipin); Bernardinelli-Seip congenital lipodystrophy 2 homolog; Bernardinelli-Seip congenital lipodystrophy type 2 protein; Bernardinelli-Seip congenital lipodystrophy type 2 protein homolog; bscl2; bscl2 {ECO:0000250; BSCL2, seipin lipid droplet biogenesis associated; G protein gamma 3 linked; Gng3lg; HMN5; MNCb-2630; PELD; seipin; SPG17; UniProtKB:Q96G97} | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
14705, 26580 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.45 mg/mL | |
PBS with 0.09% sodium azide | |
Q96G97, Q9Z2E9 | |
BSCL2 | |
KLH conjugated synthetic peptide between 78-107 amino acids from the N-terminal region of human BSCL2 | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
Ig |