Learn More
Bombesin Receptor 3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532679
Code : PN
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%); Monkey (95%); Bat, Panda, Rabbit (80%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
Bombesin Receptor 3 Polyclonal antibody specifically detects Bombesin Receptor 3 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
Bombesin Receptor 3 | |
Polyclonal | |
Unconjugated | |
BRS3 | |
BB3; BB3R; bombesin like receptor 3; bombesin receptor subtype 3; bombesin receptor subtype-3; bombesin-like receptor 3; BRS 3; BRS3; BRS-3; G-protein coupled receptor; RP1-196E23.3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
680 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P32247 | |
BRS3 | |
Synthetic 20 amino acid peptide from 2nd extracellular domain of human BRS3. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |