missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BrdU Monoclonal Antibody (BU20A), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA512502
Code : LE
Additional Details : Weight : 0.01000kg
Description
MA5-12502 targets Bromodeoxyuridine in IHC (P) applications and shows reactivity with All samples. The MA5-12502 immunogen is brdU conjugated to BSA.
BrdU Monoclonal antibody specifically detects BrdU in Chemical samples. It is validated for Immunohistochemistry (Paraffin).Specifications
BrdU | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Mouse | |
Protein G | |
RUO | |
4° C | |
Liquid | |
Chemical |
Immunohistochemistry (Paraffin) | |
BU20A | |
Unconjugated | |
5-Bromo-2-deoxyuridine; Bromodeoxyuridine | |
BrdU conjugated to BSA | |
500 μL | |
Primary | |
Antibody | |
IgG1 κ |