missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Bovine IgG (H+L) Secondary Antibody, Invitrogen™
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen A24442
Code : NEW
Additional Details : Weight : 0.01000kg
Description
The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).
Bovine IgG (H+L) Polyclonal antibody specifically detects Bovine IgG (H+L) in Bovine samples. It is validated for ELISA.Specifications
Bovine IgG (H+L) | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity Chromatography | |
RUO | |
Bovine | |
Liquid |
ELISA | |
> 4.5 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Bovine IgG (H+L) | |
2 mg | |
Secondary | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |