Learn More
BoNT-C1 Heavy Chain Mouse anti-C. botulinum, Clone: 580223, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB5425
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13440946
  • £277.83 / 100µg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
BoNT-C1 Heavy Chain Monoclonal antibody MAB5425 Specifically detects BoNT-C1 Heavy Chain in C. botulinum samples. It is validated for Immunoprecipitation, Western Blotting
Specifications
Specifications
BoNT-C1 Heavy Chain
580223
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects C. botulinum BoNT-C1 Heavy Chain in direct ELISAs. In direct ELISAs, no cross-reactivity with the Light Chains of BoNT-A, -B, -C1, -D, -E, -F, -G, or Heavy Chains of BoNT-D or -G is observed.
C. botulinum
Western Blot, Immunoprecipitation
LYOPH
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
P18640
CbC4_4096
E. coli-derived recombinant C. botulinum BoNT-C1 Heavy Chain Asn866-Glu1291 Accession # P18640
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10523893
Sterile PBS to a final concentration of 0.5 mg/mL.