Learn More
BOC Mouse anti-Human, Mouse, Clone: 273729, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB20361
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13469115
  • £286.65 / 100µg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
BOC Monoclonal antibody MAB20361 Specifically detects BOC in Human, Mouse samples. It is validated for Western Blotting
Specifications
Specifications
BOC
273729
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
BOC
Mouse myeloma cell line NS0-derived recombinant human BOC Asp31-Asp852 Accession # Q9BWV1
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
91653
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
Q9BWV1
Boc homolog (mouse), brother of CDO, Protein BOC
Mouse
IgG2a
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human and mouse BOC in direct ELISAs and Western blots.
Human, Mouse