Learn More
BMP-8 Mouse anti-Human, Clone: 158708, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1073
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13488305
  • £367.00 / 500µg
Estimated Shipment
05-07-2018
Add to basket
Description
Description
BMP-8 Monoclonal antibody MAB1073 Specifically detects BMP-8 in Human samples. It is validated for Western Blotting
Specifications
Specifications
BMP-8
Unconjugated
Murine
IgG2b
500μg
Primary
Human
158708
Purified
E.Coli-derived recombinant human BMP‑8. Ala264-His402
Protein A or G purified from hybridoma culture supernatant