Learn More
BMP-7 Monoclonal Antibody (2D6), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA518489
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide Sequence: MHVRSLRAAA PHSFVALWAP LFLLRSALAD FSLDNEVHSS FIHRRLRSQE RREMQREILS ILGLPHRPRP HLQGKHNSAP MFMLDLYNAM AVEEGGGPGG QGFSYPYKAV FSTQGPPLAS LQDSHFLTDA DMVMSFVNLV EHDKEFFHPR YHHREFRFDL SKIPEGEAVT AAEFRIYKDY IRERFDNETF RISVYQVLQE HLGRESDLFL LDSRTLWASE EGWLVFDITA TSNHWVVNPR HNLGLQLSVE TLDGQSINPK LAGLIGRHGP QNKQPFMVAF FKATEVHFRS IRSTGSKQRS QNRSKTPKNQ EALRMANVAE NSSSDQRQAC KKHELYVSFR DLGWQDWIIA PEGYAAYYCE GECAFPLNSY MNATNHAIVQ TLVHFINPET VPKPCCAPTQ LNAISVLYFD DSSNVILKKY RNMVVRACGC H
BMP-7 Monoclonal antibody specifically detects BMP-7 in Human samples. It is validated for ELISA, Western Blot.Specifications
BMP-7 | |
Monoclonal | |
0.3 mg/mL | |
PBS with no preservative; pH 7.4 | |
P18075 | |
Bmp7 | |
BMP7 (AAH08584, 1 a.a. ∽ 431 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
ELISA, Western Blot | |
2D6 | |
Unconjugated | |
Bmp7 | |
BMP; BMP7; BMP-7; bone moorphogenic protein-7; Bone morphogenetic protein; bone morphogenetic protein 7; bone morphogenetic protein 7 (osteogenic protein 1); bone morphogenetic protein 7 preproprotein; bone morphogenic protein 7; Eptotermin alfa; OP1; OP-1; Osteogenic protein 1; RP11-560A15.5 | |
Mouse | |
Affinity chromatography | |
RUO | |
655 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |