missing translation for 'onlineSavingsMsg'
Learn More

BMP-3 Mouse anti-Human, Clone: 203706, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1876

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448995

  • £420.32 / 500µg

Estimated Shipment date
from Supplier 06-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BMP-3 Monoclonal antibody MAB1876 Specifically detects BMP-3 in Human samples. It is validated for Western Blotting
Specifications

Specifications

BMP-3
203706
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
BMP3
E. coli-derived recombinant human BMP-3 Gln363-Arg472 Accession # P12645.2
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
651
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
P12645.2
BMP3, BMP-3, BMP3A, BMP-3A, bone morphogenetic protein 3, bone morphogenetic protein 3 (osteogenic), Bone morphogenetic protein 3A, bone morphogenetic protein-3, osteogenin
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human BMP-3 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows approximately 40% cross-reactivity with recombinant human (rh) BMP-3b/GDF-10. In direct ELISAs and Western blots, this antibody does not cross-react with rhBMP-2, -4, -5, -6, -7, or -8.
Human
Videos
Documents

For Research Use Only