missing translation for 'onlineSavingsMsg'
Learn More

BMP-2/BMP-4 Mouse anti-Zebrafish, Clone: 169125, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1128

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13458375

  • £337.93 / 500µg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

BMP-2/BMP-4 Monoclonal antibody MAB1128 Specifically detects BMP-2/BMP-4 in Zebrafish samples. It is validated for Western Blotting
Specifications

Specifications

BMP-2/BMP-4
169125
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
BMP2
E. coli-derived recombinant zebrafish BMP-4 Ser288-Arg400 Accession # NP_571417
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
650
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
NP_571417
BDA2, BMP2, BMP-2, BMP2A, BMP-2A, BMP2ABone morphogenetic protein 2A, BMP4, bone morphogenetic protein 2
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects zebrafish BMP-2 and BMP-4 in direct ELISAs and Western blots.
Zebrafish
Videos

For Research Use Only