missing translation for 'onlineSavingsMsg'
Learn More

BLMH, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA535295

Code : NEW

Additional Details : Weight : 0.01000kg

Product Code. 15387704

  • £325.00 / 400µL


Estimated Shipment
07-06-2018
Add to basket

Description

Description

This antibody is predicted to react with rabbit based on sequence homology.
Specifications

Specifications

BLMH
Polyclonal
PBS with 0.09% sodium azide
Q13867, Q8R016, P70645
BLMH;Bleomycin hydrolase
Rabbit
IgG
400μL
Antibody
Polyclonal
HumanþMurineþRat
Western Blot
Unconjugated
BLMH
Purified
BLMH
KLH conjugated synthetic peptide between 212-242 amino acids from the central region of human BLMH
Antigen affinity chromatography
Store at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
642
Videos