missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Blooms Syndrome Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA527384
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Jurkat, Raji, K562, NCI-H929, NIH-3T3, JC, BCL-1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Blooms Syndrome Polyclonal antibody specifically detects Blooms Syndrome in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
Blooms Syndrome | |
Polyclonal | |
Unconjugated | |
BLM | |
Blm; Bloom syndrome homolog; Bloom syndrome protein; Bloom syndrome protein homolog; Bloom syndrome RecQ like helicase; Bloom syndrome, RecQ helicase-like; BS; DNA helicase, RecQ-like type 2; mBLM; MGC126616; MGC131618; MGC131620; RecQ helicase homolog; recQ protein-like 3; RecQ2; RECQL2; RECQL3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
641 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1.34 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P54132 | |
BLM | |
Recombinant fragment corresponding to a region within amino acids 1235 and 1417 of Human BLM | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |