missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B3GNT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA521988
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, mouse stomach. Predicted reactivity: Mouse (81%), Rat (80%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
B3GNT3 Polyclonal antibody specifically detects B3GNT3 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
B3GNT3 | |
Polyclonal | |
Unconjugated | |
B3gnt3 | |
2210008L19Rik; B3GALT8; B3GAL-T8; B3gnt3; B3GN-T3; B3GNT-3; Beta-1,3-galactosyl-O-glycosyl-glycoprotein beta-1,3-N-acetylglucosaminyltransferase; Beta-1,3-galactosyltransferase 8; Beta-1,3-GalTase 8; beta-1,3-Gn-T3; beta-1,3-N-acetylglucosaminyltransferase 3; beta-1,3-N-acetylglucosaminyltransferase bGnT-3; beta3GalT8; beta3Gn-T3; beta-3-Gx-T8; BGnT-3; core 1 beta-1,3-N-acetylglucosaminyltransferase; core 1 extending beta-1,3-N-acetylglucosaminyltransferase; Core1-beta3GlcNAcT; HP10328; N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase 3; putative type II membrane protein; TMEM3; Transmembrane protein 3; UDP-Gal:beta-GlcNAc beta-1,3-galactosyltransferase 8; UDP-galactose:beta-N-acetylglucosamine beta-1,3-galactosyltransferase 8; UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 3; UNQ637/PRO1266 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10331, 72297 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q5JCS9, Q9Y2A9 | |
B3gnt3 | |
Recombinant fragment corresponding to a region within amino acids 122 and 338 of B3GNT3 (Uniprot ID#Q9Y2A9) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |