missing translation for 'onlineSavingsMsg'
Learn More

NGF, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA118378

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11552022

  • £292.00 / 20µg


Estimated Shipment
25-06-2018
Add to basket

Description

Description

By ELISA, this antibody exhibits a cross-reactivity of less than 1% to recombinant human BDNF, NT3 and NT4/5. It will not react with bovine NGF.Use at 1-3μg/ml for inhibition of biological activity in vitro. Use neat for in vivo studies at 2-10μg/ml (ED50). This antiserum completely inhibits neuronal survival and the outgrowth actions of murine NGF in chicken DRG in vitro.Reconstitute with 20 ul of distilled water. After reconstitution keep aliquots at -20ºC for a higher stability, and at 4ºC with an appropriate antibacterial agent.
Specifications

Specifications

Nerve Growth Factor beta
Polyclonal
Unconjugated
NGF
Purified
NGF
Native mouse beta NGF purified from submaxillary salivary gland.
Affinity chromatography
store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.
Primary
4803
ELISAþImmunohistochemistryþInhibition AssaysþWestern Blot
1mg/mL
Antibody with no preservative
P01138, P01139, P25427
beta Nerve Growth Factor
Rabbit
IgG
20μg
Antibody
Polyclonal
AvianþHumanþMurineþRat
Videos