Learn More
beta IG-H3 Mouse anti-Human, Clone: 348506, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB2935
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439795
  • £364.33 / 500µg
Estimated Shipment date
from Supplier 27-09-2019
Description
Description
beta IG-H3 Monoclonal antibody MAB2935 Specifically detects beta IG-H3 in Human samples. It is validated for ELISA Capture, Western Blotting
Specifications
Specifications
Beta Ig-h3/TGFBI
348506
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Q15582
beta IGH3, Beta ig-h3, BIGH3EBMD, CDB1, CDG2, CDGG1transforming growth factor, beta-induced, 68kD, CSD, CSD1, CSD2, CSD3, Kerato-epithelin, LCD1, RGD-CAP, RGD-containing collagen-associated protein, TGFBI, TGFBIP, transforming growth factor, beta-induced, 68kDa, transforming growth factor-beta-induced protein ig-h3
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human beta IG-H3 in ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant mouse beta IG-H3.
Human
Western Blot, ELISA
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TGFBI
Mouse myeloma cell line NS0-derived recombinant human beta IG-H3 Gly24-His683 Accession # Q15582
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
7045
Reconstitute at 0.5 mg/mL in sterile PBS.