Learn More
GBA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522326
Code : LD
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (84%), Rat (85%), Pig (90%), Chimpanzee (100%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GBA Polyclonal antibody specifically detects GBA in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
GBA | |
Polyclonal | |
Unconjugated | |
GBA | |
acid beta glucosidase; acid beta-glucosidase; Alglucerase; betaGC; beta-GC; Beta-glucocerebrosidase; Cholesterol glucosyltransferase; Cholesteryl-beta-glucosidase; D-glucosyl-N-acylsphingosine glucohydrolase; Gba; GBA1; GBAP; GBAP1; GC; GCase; GCB; GLUC; glucocerebrosidase; glucosidase, beta, acid; glucosylceramidase; glucosylceramidase beta; glucosylceramidase beta pseudogene 1; glucosylceramidase-like protein; imiglucerase; Lysosomal acid GCase; Lysosomal acid glucosylceramidase; lysosomal glucocerebrosidase; SGTase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2629 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.78 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P04062 | |
GBA | |
Recombinant fragment corresponding to a region within amino acids 65 and 384 of GBA (Uniprot ID#P04062) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |