missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Catenin Mouse anti-Human, Mouse, Rat, Clone: 196621, R&D Systems™
Mouse Monoclonal Antibody has been used in 4 publications
POA
Specifications
Antigen | beta-Catenin |
---|---|
Clone | 196621 |
Immunogen | E. coli-derived recombinant human beta-Catenin Ala2-Leu781 Accession # P35222 |
Host Species | Mouse |
Primary or Secondary | Primary |
Description
beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
beta-Catenin | |
E. coli-derived recombinant human beta-Catenin Ala2-Leu781 Accession # P35222 | |
Primary | |
Human, Mouse, Rat | |
Unconjugated | |
1499 | |
Purified | |
Western Blot, Immunohistochemistry | |
RUO |
196621 | |
Mouse | |
Monoclonal | |
IgG2b | |
bCatenin, beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, CTNNB1, DKFZp686D02253, FLJ25606, FLJ37923 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |