missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Actin Loading Control Monoclonal Antibody (BA3R), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515739
230.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : A0
Additional Details : Weight : 1.24740kg
Description
Storage and handling: add 0.05% sodium azide if desired. For long term storage, store at -20oC, avoiding freeze/thaw cycles.
beta Actin Loading Control Monoclonal antibody specifically detects beta Actin Loading Control in Chicken, Human, Mouse, Rabbit, Rat samples. It is validated for ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
beta Actin Loading Control | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P29751, P60706, P60709, P60710, P60711 | |
ACTB | |
Beta-actin N-terminal peptide. | |
100 μg | |
Primary | |
Chicken, Human, Mouse, Rabbit, Rat | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
BA3R | |
Unconjugated | |
ACTB | |
0610041G09Rik; AA959943; AAT6; ACT; Act4; Act-4; ACT-5; ACTA; acta1; acta1b; ACTA2; Acta-2; ACTA3; actb; actb.L; actb1; actba; ACTC; actc1; Actc-1; actc1b; ACTE; Actg; ACTG1; Actg2; ACTGE; actin; actin alpha 1; actin alpha 1, skeletal muscle; actin alpha 1, skeletal muscle b; actin alpha 2, smooth muscle; actin alpha cardiac; actin alpha cardiac 1; actin alpha cardiac muscle 1; actin alpha cardiac muscle 1b; actin beta; actin gamma 1; actin gamma 2, smooth muscle; actin, alpha 1, skeletal muscle; actin, alpha 1b, skeletal muscle; actin, alpha 2, smooth muscle, aorta; Actin, alpha cardiac muscle 1; Actin, alpha cardiac muscle 1, intermediate form; actin, alpha cardiac muscle 1b; actin, alpha skeletal muscle; Actin, alpha skeletal muscle, intermediate form; actin, alpha, cardiac 1; actin, alpha, cardiac muscle; actin, alpha, cardiac muscle 1; actin, alpha, vascular smooth muscle; actin, aortic smooth muscle; Actin, aortic smooth muscle, intermediate form; actin, beta; actin, beta 1; acti | |
Mouse | |
Protein A | |
RUO | |
100009272, 11461, 396526, 60, 81822 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |