Learn More
beta-2 Microglobulin Monoclonal Antibody (YTH470.5), PE, Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA517686
144.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
MA5-17686 targets Beta 2-microglobulin in FACS and WB applications and shows reactivity with Human samples. The MA5-17686 immunogen is rat monoclonal antibody raised against human lymphocytes.
beta-2 Microglobulin Monoclonal antibody specifically detects beta-2 Microglobulin in Human samples. It is validated for Flow Cytometry.Specifications
beta-2 Microglobulin | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P61769 | |
B2m | |
Rat monoclonal antibody raised against human lymphocytes. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry | |
YTH470.5 | |
PE | |
B2m | |
B2m; B2-microglobulin; b2m-W01; b2m-W03; b2m-Z01; b2m-Z02; b2m-Z03; Beta 2 microglobin; Beta 2 microglobulin; beta 2-microglobulin; beta chain of MHC class I molecules; Beta2 microglobulin; beta-2 microglobulin; beta-2 microglobulin preprotein; beta2m; beta2-m; beta-2-microglobin; beta-2-microglobulin; beta-2-microglobulin C-terminal fragment (55 AA); Beta-2-microglobulin form pI 5.3; B-F; CDABP0092; HDCMA22P; IMD43; Lactollin; Ly-m11; MHC Class I; microglobulin beta-2; Orla-B2m | |
Rat | |
Protein G | |
RUO | |
567 | |
4° C, do not freeze | |
Liquid |
Safety and Handling
- beta-2 Microglobulin Monoclonal Antibody (YTH470.5), PE
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Lithium dodecyl sulfate, Lithium chloride, BRIJ 35