Learn More
Pro BDNF Mouse anti-Human, Clone: 584421, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB3175
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13439935
  • £288.60 / 100µg
Estimated Shipment date
from Supplier 06-08-2020
Description
Description
Pro BDNF Monoclonal antibody MAB3175 Specifically detects Pro BDNF in Human samples. It is validated for Immunohistochemistry
Specifications
Specifications
BDNF
584421
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
BDNF
Chinese hamster ovary cell line CHO-derived recombinant human Pro-BDNF Ala19-Arg247 Accession # P23560
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
627
Sterile PBS to a final concentration of 0.5 mg/mL.
Immunohistochemistry
LYOPH
Immunohistochemistry 8-25 ug/mL
P23560
abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin
Mouse
IgG1
100 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human Pro-BDNF in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) BDNF (mature), rh beta-NGF, rhNT-3, rhNT-4, rhPro-NT-3 or rhPro-NT-4 is observed.