missing translation for 'onlineSavingsMsg'
Learn More

BDNF Polyclonal Antibody, Invitrogen™

BDNF Antibody, PA1-18375, from Invitrogen™. Species Reactivity: Human, Mouse, Rat; Applications: Western Blot.

Brand:  Invitrogen PA118375

Code : LD

Additional Details : Weight : 0.01000kg

Product Code. 11532022

  • £349.00 / 100┬ÁL


Estimated Shipment
20-06-2018
Add to basket

Description

Description

plays a role in the development of hippocampal long term potentiation; involved in regulation of synaptic plasticity.
Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: Synthetic peptide corresponding to residues H(129) S D P A R R G E L S V C D S I S E W V(148) of human BDNF protein.
Formulation: 100μL unpurified antibody in whole serum and no preservative
Synonyms: Brain-derived neurotrophic factor, Abrineurin
Uniprot Accession: P21237, P23363, P23560
Entrez GeneID: 12064, 24225, 627
Gene Symbol: BDNF
PA1-18375 detects BDNF from rat, human, and mouse samples. PA1-18375 has been successfully used in Western blot procedures. The PA1-18375 immunogen is a synthetic peptide corresponding to residues H(129) S D P A R R G E L S V C D S I S E W V(148) of human BDNF protein, conjugated to KLH. Reconstitute with 100 ul of distilled water.
Specifications

Specifications

BDNF
Unconjugated
P21237, P23363, P23560
Brain-derived neurotrophic factor, Abrineurin
Rabbit
IgG
Avoid Freeze/Thaw Cycles, store at -20°C
Polyclonal
Rat
Western Blot
100μL unpurified antibody in whole serum and no preservative
Serum
BDNF
Synthetic peptide corresponding to residues H(129) S D P A R R G E L S V C D S I S E W V(148) of human BDNF protein.
100μL
Primary
12064, 24225, 627
Videos