missing translation for 'onlineSavingsMsg'
Learn More

BAFF/BLyS/TNFSF13B Mouse anti-Human, Clone: 137314, R&D Systems™

Mouse Monoclonal Antibody

Brand:  RD Systems (Bio-Techne) MAB1241

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13448485

  • £420.32 / 500µg

Estimated Shipment date
from Supplier 09-07-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BAFF/BLyS/TNFSF13B Monoclonal antibody MAB1241 Specifically detects BAFF/BLyS/TNFSF13B in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Western Blotting
Specifications

Specifications

BAFF/BLyS/TNFSF13B
137314
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
TNFSF13B
E. coli-derived recombinant human BAFF/BLyS/TNFSF13B Ala81-Leu285 Accession # Q9Y275
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
10673
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot, Flow Cytometry, CyTOF
LYOPH
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Q9Y275
ApoL related ligand TALL-1, B lymphocyte stimulator, BAFFB-cell activating factor, B-cell-activating factor, BLyS, BLYSB-lymphocyte stimulator, CD257, CD257 antigen, Dendritic cell-derived TNF-like molecule, DTL, TALL1, TALL-1delta BAFF, TALL1Delta4 BAFF, THANK, TNF and ApoL-related leukocyte expressed ligand 1, TNF- and APOL-related leukocyte expressed ligand 1, TNF homolog that activates apoptosis, TNFSF13B, TNFSF20, tumor necrosis factor (ligand) superfamily, member 13b, tumor necrosis factor (ligand) superfamily, member 20, tumor necrosis factor ligand superfamily member 13B, tumor necrosis factor superfamily, member 13b, tumor necrosis factor-like protein ZTNF4, ZTNF4
Mouse
IgG1
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects human BAFF/BLyS/TNFSF13B in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant human (rh) TL1A/TNSF15 is observed and no cross-reactivity with rhAPRIL, rhTNF-alpha , rhFas Ligand, rhGITR Ligand, rhLIGHT, rhTRAIL, rhTRANCE, or rhTWEAK is observed.
Human
Videos
Documents

For Research Use Only