Learn More
Bacillus Spore Antigen Polyclonal Antibody, Biotin, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA173115
Code : LD
Additional Details : Weight : 0.01000kg
Description
The PA1-73115 antibody reacts with Bacillus species spores. The antiserum is unabsorbed and is expected to crossreact with various Bacillus species. PA1-73115 has been successfully used in immunofluorescence applications. The PA1-73115 antibody was raised against purified spores of B. cereus (ATCC #11778) + B. subtilis (ATCC #9372).
Bacillus Spore Antigen Polyclonal antibody specifically detects Bacillus Spore Antigen in Bacteria samples. It is validated for Immunocytochemistry.Specifications
Bacillus Spore Antigen | |
Polyclonal | |
Biotin | |
Anthrax Spore; Bacillus Spore | |
Purified spores of B. cereus (ATCC #11778) + B. subtilis (ATCC #9372). | |
1 mL | |
Primary | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
Immunocytochemistry | |
4-5 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Rabbit | |
purified | |
RUO | |
Bacteria | |
Antibody | |
IgG |