Learn More
B7-2/CD86 Mouse anti-Rat, Clone: 199602, R&D Systems™
Mouse Monoclonal Antibody
Brand:  RD Systems (Bio-Techne) MAB1340
Code : Z1
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 13458555
  • £337.93 / 500µg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
B7-2/CD86 Monoclonal antibody MAB1340 Specifically detects B7-2/CD86 in Rat samples. It is validated for Western Blotting
Specifications
Specifications
B7-2/CD86
199602
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Purified
CD86
Mouse myeloma cell line NS0-derived recombinant rat B7-2/CD86 Val29-Ile250 Accession # NP_064466
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
942
Reconstitute at 0.5 mg/mL in sterile PBS.
Western Blot
LYOPH
Western Blot 1 ug/mL
NP_064466
Activation B7-2 antigen, B70, B72, B7-2, B7-2 antigen, B-lymphocyte activation antigen B7-2, BU63, CD28 antigen ligand 2, CD28LG2B7-2 antigen), CD86, CD86 antigen, CD86 molecule, CTLA-4 counter-receptor B7.2, FUN-1, LAB72, MGC34413, T-lymphocyte activation antigen CD86
Mouse
IgG2b
500 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Monoclonal
Detects rat B7-2/CD86 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) B7-1, recombinant rat B7-1, rmB7-2, rmB7-H1, rmB7-H2, rmB7-H3, or rmPD-L2 is observed.
Rat