Learn More
B3GALT6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543534
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: EREQARHGDL LLLPALRDAY ENLTAKVLAM LAWLDEHVAF EFVLKADDDS Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Human: 100%; Mouse: 100%; Rat: 100%; Zebrafish: 77%
B3GALT6 Polyclonal antibody specifically detects B3GALT6 in Human samples. It is validated for Western Blot.Specifications
B3GALT6 | |
Polyclonal | |
Unconjugated | |
B3GALT6 | |
b3galt6; BB129894; Beta-1,3-galactosyltransferase 6; beta-1,3-GalTase 6; beta3GalT6; beta3Gal-T6; EDSP2; GAG GalTII; galactosyltransferase II; Galactosylxylosylprotein 3-beta-galactosyltransferase; GalTII; SEMDJL1; UDP-Gal:betaGal beta 1,3-galactosyltransferase 6; UDP-Gal:betaGal beta 1,3-galactosyltransferase polypeptide 6; UDP-Gal:betaGal beta 1,3-galactosyltransferase, polypeptide 6; UDP-Gal:betaGlcNAc beta 1,3-galactosyltransferase, polypeptide 6 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
126792 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96L58 | |
B3GALT6 | |
Synthetic peptide directed towards the N-terminal of human B3GALT6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |