missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B-Raf Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA514933
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with chicken and mouse based on sequence homology.
B-Raf Polyclonal antibody specifically detects B-Raf in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
B-Raf | |
Polyclonal | |
Unconjugated | |
BRAF | |
94 kDa B-raf protein; 9930012E13Rik; AA120551; AA387315; AA473386; Braf; B-raf; B-raf protein; B-raf protein isoform 1; B-raf protein isoform 2; B-Raf proto-oncogene serine/threonine-protein kinase; B-Raf proto-oncogene serine/threonine-protein kinase (p94); B-Raf proto-oncogene, serine/threonine kinase; Braf transforming gene; BRAF1; B-RAF1; Braf2; Braf-2; C230098H17; C87398; C-RMIL; D6Ertd631e; FLJ95109; MGC126806; murine sarcoma viral (v-raf) oncogene homolog B1; NS7; p94; Proto-oncogene B-Raf; proto-oncogene c-Rmil; RAFB1; RMIL; rmil serine/threonine-protein kinase; serine/threonine kinase; serine/threonine-protein kinase B-raf; Serine/threonine-protein kinase Rmil; v-raf murine sarcoma viral oncogene homolog B; v-raf murine sarcoma viral oncogene homolog B1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
673 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.34 mg/mL | |
PBS with 0.09% sodium azide | |
P15056 | |
BRAF | |
KLH conjugated synthetic peptide between 580-609 amino acids from human B-RAF | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |