Learn More
ATP13A2, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand:  Invitrogen PA118321
Code : LD
Additional Details : Weight : 0.01000kg
Product Code. 11521992
  • £229.40 / 100┬ÁL
Estimated Shipment
25-06-2018
Add to basket
Description
Description
PA1-18321 detects ATPase from rat samples.PA1-18321 has been successfully used in immunohistochemistry procedures.The PA1-18321 immunogen is a synthetic peptide corresponding to residues C-E(147) L H R Q E E A K Q V L R Y Y(161) of mouse ATP13A2, conjugated to KLH.Reconstitute with 100 ul of distilled water.
Specifications
Specifications
ATPase
Polyclonal
whole serum with no preservative
Serum
ATP13A2
Synthetic peptide corresponding to residues C-E(147) L H R Q E E A K Q V L R Y Y(161) of mouse ATP13A2.
100μL
Antibody
Polyclonal
Rat
Immunohistochemistry
Unconjugated
Atp13a2
Probable cation-transporting ATPase 13A2, EC 3.6.3., ATP13A2
Rabbit
IgG
store at -20°C, Avoid Freeze/Thaw Cycles
Primary
362645
Videos