Learn More
Atpalpha Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA532094
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: Drosophila lysate. Predicted reactivity: Drosophila (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Atpalpha Polyclonal antibody specifically detects Atpalpha in Drosophila, Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
Atpalpha | |
Polyclonal | |
Unconjugated | |
Atpalpha | |
(Na[+] + K[+])-ATPase alpha-subunit; (Na[+],K[+])ATPase alpha subunit; (Na[+]/k[+])-ATPase; alpha-ATPase; alpha-subunit of ATPase; Atp; Atp a; ATPa; atpalpha; ATP-alpha; Atpalpha-PA; Atpalpha-PB; Atpalpha-PC; Atpalpha-PD; Atpalpha-PE; Atpalpha-PF; Atpalpha-PG; Atpalpha-PH; Atpalpha-PI; Atpalpha-PJ; Atpalpha-PK; ATPase alpha; ATPase alpha subunit; ATPBagr; CG5670; CG5670-PA; CG5670-PB; CG5670-PC; CG5670-PD; CG5670-PE; CG5670-PF; CG5670-PG; CG5670-PH; CG5670-PI; CG5670-PJ; CG5670-PK; Da-47; Dmel\CG5670; Dmel_CG5670; dnATPase; JY; K ATPase; K[+] -ATPase; K[+]-ATPase; K[+]-ATPase alpha subunit; K[+]-ATPase alpha-subunit; K-ATPase; l(3)01164; l(3)01453; l(3)04492; l(3)j3B2; l(3)j3B7; l(3)j6A4; l(3)rM454; l(3)ry136; Na; Na pump alpha subunit; Na(+)/K(+) ATPase alpha subunit; Na, K ATPase a subunit; Na,K ATPase; Na,K ATPase alpha subunit; Na,K-ATPase; Na,K-ATPase alpha; Na,K-ATPase alpha subunit; Na,K-ATPase alpha-subunit; Na/K ATPase; Na/K-ATPase; Na/K-ATPase alpha-subunit; Na[ +], K[ +] -AT | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
48971 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P13607 | |
Atpalpha | |
Recombinant protein fragment corresponding to a region within amino acids 578 and 789 of Drosophilia Atpalpha | |
100 μL | |
Primary | |
Drosophila, Human | |
Antibody | |
IgG |