Learn More
ATP6V0A2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542156
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: INRADIPLPE GEASPPAPPL KQVLEMQEQL QKLEVELREV TKNKEKLRKN Sequence homology: Cow: 86%; Dog: 93%; Guinea Pig: 83%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 92%; Rat: 100%; Yeast: 79%; Zebrafish: 77%
ATP6V0A2 Polyclonal antibody specifically detects ATP6V0A2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
ATP6V0A2 | |
Polyclonal | |
Unconjugated | |
ATP6V0A2 | |
8430408C20Rik; A2; A2V-ATPase; AI385560; ARCL; ARCL2A; ATP6 ATP6N1D; ATP6A2; Atp6n1b; Atp6n1d; Atp6n2; ATP6V0A2; ATPase H+ transporting V0 subunit a2; ATPase, H+ transporting, lysosomal (vacuolar proton pump) non-catalytic accessory protein 2 (38kD); ATPase, H+ transporting, lysosomal V0 subunit a; ATPase, H+ transporting, lysosomal V0 subunit a2; AW489264; C76904; Cc1-3; Immune suppressor factor J6B7; ISF; J6B7; lysosomal H(+)-transporting ATPase V0 subunit a2; regeneration and tolerance factor; RTF; SHIF; Stv1; T-cell expressing clone j6; Tj6; TJ6M; TJ6s; vacuolar proton translocating ATPase 116 kDa subunit a; Vacuolar proton translocating ATPase 116 kDa subunit a isoform 2; vacuolar proton translocating ATPase 116-kDa subunit a2; vacuolar proton translocating ATPase 116-kDa subunit a2 isoform; V-ATPase 116 kDa; V-ATPase 116 kDa isoform a2; V-ATPase 116 kDa subunit a2; V-ATPase a2; VPH1; v-type proton ATPase 116 kDa subunit a; V-type proton ATPase 116 kDa subunit a isoform 2; V-type | |
Rabbit | |
Affinity Chromatography | |
RUO | |
23545 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9Y487 | |
ATP6V0A2 | |
Synthetic peptide directed towards the N-terminal of human ATP6V0A2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |