Learn More
ATP1B3, Mouse, Clone: 2F4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant ATP1B3.
Brand: Abnova H00000483-M05.100ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the family of Na+/K+ and H+/K+ ATPases beta chain proteins, and to the subfamily of Na+/K+ -ATPases. Na+/K+ -ATPase is an integral membrane protein responsible for establishing and maintaining the electrochemical gradients of Na and K ions across the plasma membrane. These gradients are essential for osmoregulation, for sodium-coupled transport of a variety of organic and inorganic molecules, and for electrical excitability of nerve and muscle. This enzyme is composed of two subunits, a large catalytic subunit (alpha) and a smaller glycoprotein subunit (beta). The beta subunit regulates, through assembly of alpha/beta heterodimers, the number of sodium pumps transported to the plasma membrane. The glycoprotein subunit of Na+/K+ -ATPase is encoded by multiple genes. This gene encodes a beta 3 subunit. This gene encodes a beta 3 subunit. A pseudogene exists for this gene, and it is located on chromosome 2. [provided by RefSeq]
Sequence: MTKNEKKSLNQSLAEWKLFIYNPTTGEFLGRTAKSWGLILLFYLVFYGFLAALFSFTMWVMLQTLNDEVPKYRDQIPSPGLMVFPKPVTALEYTFSRSDPTSYAGYIEDLKKFLKPYTLEEQKNLTVCPDGALFEQKGPVYVACQFPISLLQACSGMNDPDFGYSQGNPCILVKMNRIIGLKPEGVPRIDCVSKNEDIPNVAVYPHNGMIDLKYFPYYGKKLHVGYLQPLVAVQVSFAPNNTGKEVTVECKIDGSANLKSQDDRDKFLGRVMFKITARASpecifications
ATP1B3 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC011835 | |
ATP1B3 | |
ATP1B3 (AAH11835, 1 a.a. ∼ 279 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA | |
2F4 | |
Mouse monoclonal antibody raised against a full length recombinant ATP1B3. | |
ATP1B3 | |
ATPB-3/CD298/FLJ29027 | |
Mouse | |
Affinity chromatography | |
RUO | |
483 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |