missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP13A2 Polyclonal Antibody, DyLight™ 650, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA522850
Code : LD
Additional Details : Weight : 0.01000kg
Description
Suggested positive control: human and mouse brain membrane prep lysate.
ATP13A2 Polyclonal antibody specifically detects ATP13A2 in Human, Mouse samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ATP13A2 | |
Polyclonal | |
DyLight 650 | |
ATP13A2 | |
1110012E06Rik; AA589443; AT132 antibody; Atp13a2; ATPase 13A2; ATPase type 13A2; Cation-transporting ATPase 13A2; CLN12; HSA9947; HSA9947 antibody; KRPPD; PARK9; probable cation-transporting ATPase 13A2; putative ATPase; RGD1307977; RP1-37C10.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23400, 74772 | |
4° C, store in dark | |
Liquid |
Immunohistochemistry, Western Blot | |
0.75 mg/mL | |
50mM sodium borate with no preservative | |
Q9CTG6, Q9NQ11 | |
ATP13A2 | |
A synthetic peptide within the internal region of residues 200-300 of the human ATP13A2 protein. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |