missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG4A Polyclonal Antibody, Invitrogen™
Sheep Polyclonal Antibody
Brand: Invitrogen OSA00040W
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 150 μL of distilled water.
Autophagy is the process by which endogenous proteins and damaged organelles are destroyed intracellularly. Autophagy is postulated to be essential for cell homeostasis and cell remodeling during differentiation, metamorphosis, non-apoptotic cell death, and aging. Reduced levels of autophagy have been described in some maligt tumors, and a role for autophagy in controlling the unregulated cell growth linked to cancer has been proposed. This gene encodes a member of the autophagin protein family. The encoded protein is also designated as a member of the C-54 family of cysteine proteases. Transcript variants that encode distinct isoforms have been identified.Specifications
ATG4A | |
Polyclonal | |
ATG4A | |
Cysteine protease ATG4A, autophagin-2, Apg4a, AUTL2 | |
Sheep | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q8WYN0, Q8C9S8 | |
ATG4A | |
A synthetic peptide from human, mouse cysteine protease ATG4A | |
RUO | |
115201, 666468 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |