missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSA00027W
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 150 μL of distilled water.
APG12 is required for autophagy. APG5 conjugates to ATG12 and associates with isolation membrane to form cup-shaped isolation membrane and autophagosome. The conjugate detaches from the membrane immediately before or after autophagosome formation is completed.Specifications
ATG12 | |
Polyclonal | |
Atg12 | |
APG12, APG12L, FBR93, HAPG12 | |
Rabbit | |
150 μL | |
Primary | |
Drosophila | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
Q9VTU1 | |
ATG12 | |
A synthetic peptide from aa region 20-70 of drosophila ATG12 conjugated to an immunogenic carrier protein has been used as the antigen | |
RUO | |
39383 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |