missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen OSU00003W
Code : LD
Additional Details : Weight : 0.01000kg
Description
Reconstitute with 150 μL of distilled water.
Involved in axon growth. Plays an essential role in neurite extension of cerebellar granule cells. Interacts with GABARAP and GABARAPL2.Specifications
ULK1 | |
Polyclonal | |
ULK1 | |
Serine/threonine-protein kinase ULK1, ULK1, UNC51, Unc51.1, KIAA0722, FLJ38455 | |
Rabbit | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Immunohistochemistry, Western Blot | |
Unconjugated | |
O75385, O70405 | |
ULK1 | |
A synthetic peptide from aa region 350-410 of human ULK1 conjugated to an immunogenic carrier protein has been used as the antigen | |
RUO | |
22241, 360827, 8408 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Lyophilized |